Fitting, Polypropylene, Straight, Hosebarb to Thread Adapter, 1/16" ID x 10-32 UNF(M)
Catalog # MFLX51517-03
Supplier: Avantor Fluid Handling
About this item
No tools needed for assembly.
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimise downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Barbed fittings are designed for quick and easy assembly without tools. Combine these simple and economical fittings with our flexible tubing for use in your low-pressure applications (generally less than 30 psi).
- Taille de l'ardillon/Tube Ø int. (port 2):1/16"
- Type de connection:Thread to Barb
- Flow pattern:Straight
- Désignation:Fitting, Polypropylene, Straight, Hosebarb to Thread Adapter, 1/16" ID x 10-32 UNF(M)
- Seq:33
- Environmentally Preferable:
- Modèle:51517-03
- Cdt:1
- Fitting material:Polypropylene
- Thread size UNF(M) (port 2):10-32"
Frequently Bought Together