Masterflex® Fitting, Black PVDF, One-Way Stopcock, Male Luer Lock to Male Luer Lock; 10/PK
Référence MFLX31200-90
Fournisseur: Avantor Fluid Handling
À propos de cet article
Build leakproof fluid handling systems.
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimise downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
- Designed for quick and easy assembly with flexible tubing
These one-way stopcocks feature an all-black PVDF construction.
- Flow pattern:One-Way
- Désignation:Luer stopcock, 1-way, male luer lock x male luer lock, black PVDF, pack of 10
- Seq:02
- Environmentally Preferable:
- Modèle:31200-90
- Wetted materials:Black PVDF
- Cdt:10
- Inlet port:Male Luer Lock
- Outlet port:Male Luer Lock
Frequently Bought Together