Masterflex® Fitting, Stainless Steel, Y-Connector, Hose Barb Union, 3/8" ID; 1/EA
N° de catalogue MFLX30620-24
Fournisseur: Avantor Fluid Handling
À propos de cet article
Ensure a tight seal every time.
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimise downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
- Designed for quick and easy assembly with flexible tubing
- Connection Type: 3/8" ID; 1/EA
Combine these simple, economical fittings with flexible tubing for use in your low pressure applications.
Ideal for applications such as pharmaceuticals, laboratory, industrial/manufacturing, water sampling, food processing.
- Taille de l'ardillon/Tube Ø int. (port 1):3/8"
- Taille de l'ardillon/Tube Ø int. (port 2):3/8"
- Type de connection:Barbed Union
- Flow pattern:Y
- Désignation:Fitting, Stainless Steel, Y-Connector, Hose Barb Union, 3/8" ID; 1/EA
- Seq:04
- Environmentally Preferable:
- Modèle:30620-24
- Sterilisation:Autoclave, ethylene oxide (ETO), or gamma
- Garantie:1 Month
- Cdt:1
- Barb size/tubing int.Ø (port 3):3/8"
- Fitting material:Stainless Steel 316
Frequently Bought Together