Masterflex® Fitting, Polycarbonate, Three Ports, Manifolds with Female Luer Locks, 360° Rotation, Non-Sterile; 10/PK
N° de catalogue MFLX30600-41
Fournisseur: Avantor Fluid Handling
Spécifications
Voir toutes les configurations du produit- Wetted materials:
- Polycarbonate
- Polycarbonate
- Polycarbonate
- Polycarbonate
- Garantie:
- Out of box
- Out of box
- Out of box
- Affichage actuel
Spécifications
À propos de cet article
Regulate flow without any leaking.
- Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimise downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Polycarbonate manifolds have polyurethane stems and HDPE handles. Ports have female luer locks.
- Flow pattern:3 Ports, 360 Degree Rotation
- Désignation:Fitting, Polycarbonate, Three Ports, Manifolds with Female Luer Locks, 360° Rotation, Non-Sterile; 10/PK
- Environmentally Preferable:
- Wetted materials:Polycarbonate
- Garantie:Out of box
- Cdt:10
- Inlet port:Female Luer
- Outlet port:Female Luer
Frequently Bought Together