Masterflex® L/S® Easy-Load® Pump Head, Replacement Rotor and Bearing Assembly for 07518-60, PPS Housing, SS Rotor
Catalog # MFLX07518-68
Supplier: Avantor Fluid Handling
About this item
Get the most our of your Masterflex® Pump Head with these replacement parts.
- Choose replacement parts suitable for your L/S® and I/P® pump head
- Suitable for Easy-Load® Pump Head 07518-60
Replacement parts to keep your pump head operating even longer. Two end bell assemblies are required for a complete head assembly.
- Pump series:L/S
- Désignation:Easy-Load® Pump Head, Replacement Rotor and Bearing Assembly, PPS Housing, SS Rotor; 1/Ea
- Seq:41
- Environmentally Preferable:
- Modèle:07518-68
- Open-head sensor:No
- Cdt:1
- Matériau du boîtier:PPS
- Rotor material:Stainless Steel
Frequently Bought Together