Masterflex® L/S® Standard Pump Head, Mounting Hardware, Mounts One L/S 36 Pump Head, Stainless Steel; 2/Set
Référence MFLX07036-01
Fournisseur: Avantor Fluid Handling
À propos de cet article
Get the most our of your Masterflex® Pump Head with these replacement parts.
- Choose replacement parts suitable for your L/S® and I/P® pump head
- Suitable for use with L/S® Standard Pump Heads 07036-30 and -31
Replacement parts to keep your pump head operating even longer. Two end bell assemblies are required for a complete head assembly.
- Nombre de canaux:1
- Pump series:L/S
- Désignation:Standard Pump Head, Mounting Hardware, Mounts One L/S® 36 Pump Head, Stainless Steel; 2/Set
- Seq:33
- Environmentally Preferable:
- Modèle:07036-01
- Open-head sensor:No
- Tubing sizes accepted:L/S 36
- Garantie:1 year
- Cdt:1
Frequently Bought Together