Masterflex® I/P® Standard Pump Head, Replacement Rotor Assembly, Long Shaft, SS Rotor
Référence MFLX07019-83
Fournisseur: Avantor Fluid Handling
À propos de cet article
Get the most our of your Masterflex® Pump Head with these replacement parts.
- Choose replacement parts suitable for your L/S® and I/P® pump head
- Suitable for use with I/P® Standard pump heads 07019-01, -41, -26, -51.
Replacement parts to keep your pump head operating even longer. Two end bell assemblies are required for a complete head assembly.
- Pump series:I/P
- Désignation:Masterflex I/P® Standard Pump Head, Replacement Rotor Assembly, Long Shaft, SS Rotor; 1/Ea
- Seq:27
- Environmentally Preferable:
- Modèle:07019-83
- Open-head sensor:No
- Tubing sizes accepted:I/P 73, 82
- Garantie:2 years
- Cdt:1
- Rotor material:Stainless Steel
Frequently Bought Together