Masterflex® L/S® Standard Pump Head, Replacement Rotor Assembly, Precision Tubing Sizes, SS
Catalog # MFLX07013-95
Supplier: Avantor Fluid Handling
About this item
Get the most our of your Masterflex® Pump Head with these replacement parts.
- Choose replacement parts suitable for your L/S® and I/P® pump head
- Suitable for use with L/S® Standard Pump Heads 07013-21 and -52; 07014-21 and -52; 07016-21 and -52; 07017-21 and -52; 07018-21 and -52
Replacement parts to keep your pump head operating even longer. Two end bell assemblies are required for a complete head assembly.
- Pump series:L/S
- Désignation:Standard Pump Head, Replacement Rotor Assembly, Precision Tubing Sizes, SS Rotor; 1/Ea
- Seq:05
- Environmentally Preferable:
- Modèle:07013-95
- Open-head sensor:No
- Tubing sizes accepted:L/S 13, 14, 16, 25, 17, 18
- Garantie:2 years
- Cdt:1
- Rotor material:Stainless Steel
Frequently Bought Together