Masterflex® Fitting, Clear Polypropylene, Elbow, Hose Barb Union, 1/16" ID; 25/PK
Catalog # MFLX06365-17
Supplier: Avantor Fluid Handling
About this item
Quick and easy assembly without tools.
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimise downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
- Connection Type: 1/16" ID
Create 90° bends in flexible tubing—without the kinks!
- Taille de l'ardillon/Tube Ø int. (port 1):¹⁄₁₆"
- Taille de l'ardillon/Tube Ø int. (port 2):¹⁄₁₆"
- Type de connection:Barbed Union
- Flow pattern:Elbow
- Désignation:Fitting, Clear Polypropylene, Elbow, Hose Barb Union, ¹⁄₁₆" ID; 25/PK
- Seq:08
- Environmentally Preferable:
- Modèle:06365-17
- Orifice dimensions:0,03125"
- Sterilisation:Autoclave or ethylene oxide (ETO)
- Cdt:25
- Fitting material:Polypropylene
Frequently Bought Together