Catalyst Pump Tubing, 2-Stop, PharMed® BPT, 1.6 mm ID; 12/Pk
N° de catalogue MFLX97100-23
Fournisseur: Avantor Fluid Handling
À propos de cet article
Start pumping with little effort - preconfigured Links for quick and easy setup.
- Four formulations offer wide range of chemical compatibility
- Save time with already assembled links
- Use with the Catalyst extenstion coils / tubing to complete your system
- FH15 and FH30 utilize the stopped assemblies that have an ID of: 0.8, 1.6, 3.2, and 4.8 mm
- FH100M (with the large cartridges) can use assemblies with an ID of: 0.8, 1.6, 3.2, 4.8, and 6.4 mm
- FH100M (with the small cartridges) can use assemblies with an ID of: 0.16, 0.25, 0.89, 1.42, 2.06, and 2.79 mm
To complete your Catalyst® pumping system, order two-stop color-coded and extension tubing. Tubing comes in various formulations for almost any pumping application.
The stops on the two-stop tubing are color-coded according to tubing ID.
Order extension tubing separately. For technical assistance selecting tubing formulations and sizes, contact our Application Specialists.
- Tube Ø int.:1,60 mm (0,063")
- Max. temperature:132 °C (270 °F)
- Couleur:brun roux
- Désignation:Stopped Tubing Assembly, PharMed BPT, 1,6 mm ID, 12 pk
- Longueur:0,4064 m
- Environmentally Preferable:
- GMP compliant:Yes
- Length per pack:4,8768 m (15,96 ft.)
- Regulatory approvals:ADCF;European Pharmacopoeia;Non-DEHP;RoHS;USP
- Sterilisation method autoclave:Yes
- Sterilisation method EtO:Yes
- Sterilisation method gamma:Yes
- Température min.:–51 °C (–60 °F)
- Series:Catalyst
- Clarity:Opaque
- Matériau de tuyaux:PharMed® BPT
- Cdt:12
Frequently Bought Together