Masterflex® Smart Gasket® Sanitary Sampling Gasket, Silicone, 2 Port, 1" Tri-Clamp; 1/EA
Référence MFLX30543-08
Fournisseur: Avantor Fluid Handling
À propos de cet article
Smart Gasket lets you take samples or readings from a sanitary line.
- Measure temperature, take a sample, or dose chemicals into your process line
- Install in between two sanitary fittings
The Smart Gasket is used as a standard sanitary gaskets with standard Tri-clamp® fittings. The gaskets have molded ports that allow either a thermocouple or sampling tubes to be inserted into the line. Order the appropriate Smart Gasket clamp separately.
- Sanitary clamp size (Port 1):1-1/2"
- Connection type:Sanitary
- Désignation:Sanitary Sampling Gasket, Silicone, 2 Port, 1" Tri-Clamp; 1/EA
- Seq:26
- Environmentally Preferable:
- Cdt:1
- Fitting material:Platinum-Cured Silicone
- Sanitary gasket size:1"
- No. of ports:2
- Garantie:1 year
Frequently Bought Together