Watch glasses
Watch glass, glass, with fused rim, 180 mm
N° de catalogue HECH40010018
Fournisseur: GLASWARENFABRIK KARL HECHT
Aller à
Product Details & Documents
Evaporating dishes, made of glass, with fused rim.
- Good chemical restistance
- Available with or without hole
Certifications: DIN 12 341
Spécifications
- Ø:180 mm
- Désignation:Watch glass, glass, with fused rim
- Seq:21