2-(4-Hydroxyphényl)ethanol ≥98%
100 g
N° de catalogue CAYM27600-100
Fournisseur: Cayman Chemical
2-(4-Hydroxyphényl)ethanol ≥98%
N° de catalogue CAYM27600-100
Fournisseur: Cayman Chemical
Numéro CAS: 501-94-0
Aller à
Product Details & Documents
Tyrosol is a phenol that has been found in olives and olive oil and has diverse biological activities.
Caution: This product is not for human or veterinary use.
Synonymes: NSC 59876, p-HPEA, Tyrosol, 4-Hydroxyphenethyl alcohol
- Numéro CAS:501-94-0
- Formule:C₈H₁₀O₂
- Numéro MDL:MFCD00002902
- Poids moléculaire:138.17 g/mol
- Température de stockage:Congélateur
Spécifications
Spécifications
- Cdt:100 g
Spécifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



