RABBIT COAGULASE PLASMA WITH EDTA 1 * 30 mL
JJBE240827
Frequently Bought Together
LETHEEN BROTH (MODIFIED) (DEHYDRATED CUL 1 * 500 g
: PanReac AppliChem
VIOLET RED BILE GLUCOSE AGAR 1 * 500 g
: LIOFILCHEM
Boîtes de contact pré-coulées, Compact Dry
: Shimadzu Diagnostics Europe
Dehydrated Media
: THERMO FISHER DIAGNOSTICS
Milieux de culture déshydratés pré-pesés en sachets
: VWR Chemicals
[EN]SWAB P/WIPE PREMOISTENED BLUE SPONGE 1 * 50 Pce
: MEDICAL WIRE EQUIPMENT
VWR®, Milieux de culture prêts à l'emploi, en flacons
: VWR Chemicals
Settle plates, gamma-irridiated, triple-bagged, ICR (isolator, cleanroom)
: MERCK LIFE SCIENCE
MyoTonic™ Serum-Free Growth 10.00 $155.00 $1,550.00 null
: MARKET SOURCE PART PROCESS