Saisie de commande
Switzerland
Lien pour commandesContactUsLinkComponent
4-Hydroxybenzoate d'éthyle 96%

4-Hydroxybenzoate d'éthyle 96%

Bottle, 500 g
Catalog # APOSOR923091-500G
Supplier:   Apollo Scientific
4-Hydroxybenzoate d'éthyle 96%
Catalog # APOSOR923091-500G
Supplier:  Apollo Scientific
CAS Number:  120-47-8
4-Hydroxybenzoate d'éthyle 96%

Bottle, 500 g

CAS Number:  120-47-8
Price
SFr. 69.30
Log in for availability
1 * 500 g
Product Details & Documents
Synonyms:  Nipagin A, Ethyl paraben, Ethyl p-hydroxybenzoate, Ethylparaben, Ethyl 4-hydroxybenzoate
  • CAS Number:120-47-8
  • Formula:C₉H₁₀O₃
  • MDL Number:MFCD00002353
  • Molecular Weight:166.18 g/mol
  • Storage Temperature:Température ambiante
Specifications

Specifications

  • Type de conditionnement:
    Bottle
  • Cdt:
    500 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.