Human sera
Human serum, HIV tested, heat inactivated, Filtré de manière stérile, Frozen
: Biowest
Human serum 'off-the-clot' is processed from human blood that has been coagulation-induced. It is collected from volunteer donors.
Mainly sourced from France, Germany, Canada or USA, each batch is rigorously controlled and screened for Hepatitis B (HBS), Hepatitis C (HCV) and HIV Types 1 and 2 (HIV 1/2).
Important note: Products of human origin should be considered potentially infectious and handled accordingly.
: Special treatments are available on request. Please contact your local Avantor Sales Office for more information.
- Format du sérum:Frozen
- Stérilité:Filtré de manière stérile
- Désignation:Human serum, HIV tested, heat inactivated