Pt100 probe, L=35 cm, M8 connector to connect to LIBERO CE, bendable once at room temperature (do not kink). Temperature range -200 °C..+200 °C. 1 * 1 Pce
Pt100 probe, L=35 cm, M8 connector to connect to LIBERO CE, bendable once at room temperature (do not kink). Temperature range -200 °C..+200 °C. 1 * 1 Pce