ContactUsLinkComponent
Pt100 probe, L=35 cm, M8 connector to connect to LIBERO CE, bendable once at room temperature (do not kink). Temperature range -200 °C..+200 °C. 1 * 1 Pce
N° de catalogue ELPR802288
Fournisseur:  ELPRO-BUCHS
Pt100 probe, L=35 cm, M8 connector to connect to LIBERO CE, bendable once at room temperature (do not kink). Temperature range -200 °C..+200 °C. 1 * 1 Pce
N° de catalogue ELPR802288
Fournisseur:  ELPRO-BUCHS
Frequently Bought Together