Recombinat Protein,Biotinylated Human Angiopoietin-2 ANGPT2 Protein, His,Avitag™,Biotinylated Human Angiopoietin-2, His,Avitag (AN2 -H82E7) is expressed from human 293 cells (HEK293). It contains AA Lys 275 - Phe 496 (Accession AAI26201.1).This prot 1 * 200 µG
N° de catalogue ACRBAN2-H82E7200UG
Fournisseur: ACROBIOSYSTEMS
Frequently Bought Together