Recombinat Protein,Human CXADR CAR Protein, His Tag,Human CXADR, His Tag (CXR-H5222) is expressed from human 293 cells (HEK293). It contains AA Leu 20 - Gly 237 (Accession NP001329.1).This protein carries a polyhistidine tag at the C-terminus. The ca 1 * 1 mg
Catalog # ACRBCXR-H5222-1MG
Supplier: ACROBIOSYSTEMS
Frequently Bought Together