ContactUsLinkComponent
MABSol® Biotinylated Human GM-CSF (GMF-H8214) is expressed from human HEK293 cells. It contains AA Ala 18 - Glu 144 (Accession NP0 00749.2) It is the biotinylated form of ActiveMax® Human GM-CSF (Cat GMF-H4214). null
Référence ACRBGMF-H8214-25UG
Fournisseur:  ACROBIOSYSTEMS
MABSol® Biotinylated Human GM-CSF (GMF-H8214) is expressed from human HEK293 cells. It contains AA Ala 18 - Glu 144 (Accession NP0 00749.2) It is the biotinylated form of ActiveMax® Human GM-CSF (Cat GMF-H4214). null
Référence ACRBGMF-H8214-25UG
Fournisseur:  ACROBIOSYSTEMS
Frequently Bought Together