ContactUsLinkComponent
Recombinat Protein,Human Annexin A5 Protein, Tag Free,Human ANXA5, Tag Free (AN5-H5118) is expressed from E.coli cells. It contains AA Met 1 - Asp 320 (Accession P08758-1).This protein carries no tag. The protein has a calculated MW of 35.9 kDa. The 1 * 100 µG
Catalog # ACRBAN5-H5118100UG
Supplier:  ACROBIOSYSTEMS
Recombinat Protein,Human Annexin A5 Protein, Tag Free,Human ANXA5, Tag Free (AN5-H5118) is expressed from E.coli cells. It contains AA Met 1 - Asp 320 (Accession P08758-1).This protein carries no tag. The protein has a calculated MW of 35.9 kDa. The 1 * 100 µG
Catalog # ACRBAN5-H5118100UG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together