ContactUsLinkComponent
Human GITR Ligand, His Tag (GIL-H5249) is expressed from human 293 cells (HEK293). It contains AA Gln 50 - Ser 177 (Accession AAH69 319.1). 1 * 1 mg
N° de catalogue ACRBGIL-H5249-1MG
Fournisseur:  ACROBIOSYSTEMS
Human GITR Ligand, His Tag (GIL-H5249) is expressed from human 293 cells (HEK293). It contains AA Gln 50 - Ser 177 (Accession AAH69 319.1). 1 * 1 mg
N° de catalogue ACRBGIL-H5249-1MG
Fournisseur:  ACROBIOSYSTEMS
Frequently Bought Together