Recombinat Protein,FITC-Labeled Human ErbB3 Her3 Protein, His Tag,FITC-Labeled Human ErbB3, His Tag (ER3-HF2H5) is expressed from h uman 293 cells (HEK293). It contains AA Ser 20 - Thr 643 (Accession P21860-1). It is the FITC labeled form of Human E 1 * 200 µG
N° de catalogue ACRBER3-HF2H5200UG
Fournisseur: ACROBIOSYSTEMS
Frequently Bought Together