ContactUsLinkComponent
FITC-Labeled Human Her2, His Tag (HE2-HF224) is expressed from human 293 cells (HEK293). It contains AA Thr 23 - Thr 652 (Accession P04626-1). It is the FITC labeled form of Human Her2, His Tag (HE2-H5225). 1 * 200 µG
N° de catalogue ACRBHE2-HF224200UG
Fournisseur:  ACROBIOSYSTEMS
FITC-Labeled Human Her2, His Tag (HE2-HF224) is expressed from human 293 cells (HEK293). It contains AA Thr 23 - Thr 652 (Accession P04626-1). It is the FITC labeled form of Human Her2, His Tag (HE2-H5225). 1 * 200 µG
N° de catalogue ACRBHE2-HF224200UG
Fournisseur:  ACROBIOSYSTEMS
Frequently Bought Together