Recombinat Protein,Human HABP1 C1QBP Protein, His Tag,Human HABP1, His Tag (C1P-H5122) is expressed from E.coli cells. It contains AA His 75 - Gln 282 (Accession AAH00435).This protein carries a polyhistidine tag at the C-terminus. The protein has a 1 * 100 µG
Référence ACRBC1P-H5122100UG
Fournisseur: ACROBIOSYSTEMS
Frequently Bought Together