ContactUsLinkComponent
FITC-Labeled Human BCMA, His Tag (Cat. No. BCA-HF2H1) is expressed from human HEK293 cells. It contains AA Met 1 - Ala 54 (Accession Q02223-1). It is the FITC labeled form of Human BCMA, His Tag (Cat. No. BCA-H522y). 1 * 25 µG
N° de catalogue ACRBBCA-HF2H1-25UG
Fournisseur:  ACROBIOSYSTEMS
FITC-Labeled Human BCMA, His Tag (Cat. No. BCA-HF2H1) is expressed from human HEK293 cells. It contains AA Met 1 - Ala 54 (Accession Q02223-1). It is the FITC labeled form of Human BCMA, His Tag (Cat. No. BCA-H522y). 1 * 25 µG
N° de catalogue ACRBBCA-HF2H1-25UG
Fournisseur:  ACROBIOSYSTEMS
Frequently Bought Together