PE-Labeled Human PD-1, Fc tag, His tag (recommended for neutralizing assay) (Cat. No. PD1-HP2F2) is produced via conjugation of PE t o Human PD-1, Fc tag, His tag under optimal conditions. Human PD-1, Fc tag, His tag is expressed in HEK293 cells cont 1 * 50 Tests
Référence ACRBPD1HP2F250TEST
Fournisseur: ACROBIOSYSTEMS
Frequently Bought Together