ContactUsLinkComponent
Biotinylated Human TSLP, His,Avitag (TSP-H82Eb) is expressed from human 293 cells (HEK293). It contains AA Tyr 29 - Gln 159 (Accessi on Q969D9-1). 1 * 200 µG
Catalog # ACRBTSP-H82EB200UG
Supplier:  ACROBIOSYSTEMS
Biotinylated Human TSLP, His,Avitag (TSP-H82Eb) is expressed from human 293 cells (HEK293). It contains AA Tyr 29 - Gln 159 (Accessi on Q969D9-1). 1 * 200 µG
Catalog # ACRBTSP-H82EB200UG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together