Recombinat Protein,Biotinylated Human BLAME SLAMF8 Protein, His,Avitag™,Biotinylated Human BLAME, His,Avitag (BLE-H82E4) is express ed from human 293 cells (HEK293). It contains AA Ala 23 - Asp 233 (Accession Q9P0V8-1).This protein carries a polyhis 1 * 25 µG
N° de catalogue ACRBBLE-H82E4-25UG
Fournisseur: ACROBIOSYSTEMS
Frequently Bought Together