ContactUsLinkComponent
FITC-Labeled Human Glypican 3, His Tag (Cat. No. GP3-HF2H1) is expressed from human HEK293 cells. It contains AA Gln 25 - His 559 (A ccession P51654-1). It is the FITC labeled form of Human Glypican 3, His Tag (Cat. No. GP3-H52H4). 1 * 200 µG
N° de catalogue ACRBGP3-HF2H1200UG
Fournisseur:  ACROBIOSYSTEMS
FITC-Labeled Human Glypican 3, His Tag (Cat. No. GP3-HF2H1) is expressed from human HEK293 cells. It contains AA Gln 25 - His 559 (A ccession P51654-1). It is the FITC labeled form of Human Glypican 3, His Tag (Cat. No. GP3-H52H4). 1 * 200 µG
N° de catalogue ACRBGP3-HF2H1200UG
Fournisseur:  ACROBIOSYSTEMS
Frequently Bought Together