ContactUsLinkComponent
FITC-Labeled Human BCMA, Fc Tag (Cat. No. BCA-HF254) is expressed from human HEK293 cells. It contains AA Met 1 - Ala 54 (Accession Q02223-1). It is the FITC labeled form of Human BCMA, Fc Tag (Cat. No. BC7-H5254). 1 * 25 µG
N° de catalogue ACRBBCA-HF254-25UG
Fournisseur:  ACROBIOSYSTEMS
FITC-Labeled Human BCMA, Fc Tag (Cat. No. BCA-HF254) is expressed from human HEK293 cells. It contains AA Met 1 - Ala 54 (Accession Q02223-1). It is the FITC labeled form of Human BCMA, Fc Tag (Cat. No. BC7-H5254). 1 * 25 µG
N° de catalogue ACRBBCA-HF254-25UG
Fournisseur:  ACROBIOSYSTEMS
Frequently Bought Together