ContactUsLinkComponent
Canine GITR, His Tag (GIR-C52H6) is expressed from human 293 cells (HEK293). It contains AA Gly 23 - Pro 154 (Accession D7F619-1). 1 * 1 mg
N° de catalogue ACRBGIR-C52H6-1MG
Fournisseur:  ACROBIOSYSTEMS
Canine GITR, His Tag (GIR-C52H6) is expressed from human 293 cells (HEK293). It contains AA Gly 23 - Pro 154 (Accession D7F619-1). 1 * 1 mg
N° de catalogue ACRBGIR-C52H6-1MG
Fournisseur:  ACROBIOSYSTEMS
Frequently Bought Together