ContactUsLinkComponent
Human GITR Ligand, Fc Tag (GIL-H526a) is expressed from human 293 cells (HEK293). It contains AA Gln 50 - Ser 177 (Accession AAH693 19.1). 1 * 1 mg
N° de catalogue ACRBGIL-H526A-1MG
Fournisseur:  ACROBIOSYSTEMS
Human GITR Ligand, Fc Tag (GIL-H526a) is expressed from human 293 cells (HEK293). It contains AA Gln 50 - Ser 177 (Accession AAH693 19.1). 1 * 1 mg
N° de catalogue ACRBGIL-H526A-1MG
Fournisseur:  ACROBIOSYSTEMS
Frequently Bought Together