ContactUsLinkComponent
Rhesus macaque CD155, His Tag (CD5-R52H5) is expressed from human 293 cells (HEK293). It contains AA Asp 28 - Asn 343 (Accession Q0 MSE6-1). 1 * 100 µG
N° de catalogue ACRBCD5-R52H5100UG
Fournisseur:  ACROBIOSYSTEMS
Rhesus macaque CD155, His Tag (CD5-R52H5) is expressed from human 293 cells (HEK293). It contains AA Asp 28 - Asn 343 (Accession Q0 MSE6-1). 1 * 100 µG
N° de catalogue ACRBCD5-R52H5100UG
Fournisseur:  ACROBIOSYSTEMS
Frequently Bought Together