ContactUsLinkComponent
Recombinat Protein,Human Serpin F1 PEDF Protein, His Tag,Human Serpin F1, His Tag (SE1-H5221) is expressed from human 293 cells (HE K293). It contains AA Gln 20 - Pro 418 (Accession NP002606).This protein carries a polyhistidine tag at the C-terminu 1 * 100 µG
N° de catalogue ACRBSE1-H5221100UG
Fournisseur:  ACROBIOSYSTEMS
Recombinat Protein,Human Serpin F1 PEDF Protein, His Tag,Human Serpin F1, His Tag (SE1-H5221) is expressed from human 293 cells (HE K293). It contains AA Gln 20 - Pro 418 (Accession NP002606).This protein carries a polyhistidine tag at the C-terminu 1 * 100 µG
N° de catalogue ACRBSE1-H5221100UG
Fournisseur:  ACROBIOSYSTEMS
Frequently Bought Together