FITC-Labeled Human Mesothelin (296-580), His Tag (Cat. No. MSN-HF223) is expressed from human HEK293 cells. It contains AA Glu 296 - Gly 580 (Accession AAH09272). It is the FITC labeled form of Human Mesothelin (296-580), His Tag (Cat. No. MSN-H522 1 * 200 µG
Catalog # ACRBMSN-HF223200UG
Supplier: ACROBIOSYSTEMS
Frequently Bought Together