ContactUsLinkComponent
Human CD58, Fc Tag (LF3-H5256) is expressed from human 293 cells (HEK293). It contains AA Phe 29 - Arg 215 (Accession AAH05930). 1 * 100 µG
  ACRBLF3-H5256100UG
Human CD58, Fc Tag (LF3-H5256) is expressed from human 293 cells (HEK293). It contains AA Phe 29 - Arg 215 (Accession AAH05930). 1 * 100 µG
  ACRBLF3-H5256100UG
Frequently Bought Together