Recombinat Protein,Human PROCR EPCR CD201 Protein, Fc Tag,Human PROCR, Fc Tag (PRR-H5254) is expressed from human 293 cells (HEK29 3). It contains AA Ser 18 - Thr 209 (Accession AAH14451).This protein carries a human IgG1 Fc tag at the C-terminus. as 1 * 100 µG
N° de catalogue ACRBPRR-H5254100UG
Fournisseur: ACROBIOSYSTEMS
Frequently Bought Together