FITC-Labeled Human CD19 (20-291), His Tag (Cat. No. CD9-HF2H2) is expressed from human HEK293 cells. It contains AA Pro 20 - Lys 291 (Accession P15391-1). It is the FITC labeled form of Human CD19, His Tag (Cat. No. CD9-H52H2). 1 * 200 µG
N° de catalogue ACRBCD9-HF2H2200UG
Fournisseur: ACROBIOSYSTEMS
Frequently Bought Together