ContactUsLinkComponent
FITC-Labeled Human CD19 (20-291), Fc Tag (Cat. No. CD9-HF251) is expressed from human HEK293 cells. It contains AA Pro 20 - Lys 291 (Accession P15391-1). It is the FITC labeled form of Human CD19 (20-291), Fc Tag (Cat. No. CD9-H5259). 1 * 25 µG
N° de catalogue ACRBCD9-HF251-25UG
Fournisseur:  ACROBIOSYSTEMS
FITC-Labeled Human CD19 (20-291), Fc Tag (Cat. No. CD9-HF251) is expressed from human HEK293 cells. It contains AA Pro 20 - Lys 291 (Accession P15391-1). It is the FITC labeled form of Human CD19 (20-291), Fc Tag (Cat. No. CD9-H5259). 1 * 25 µG
N° de catalogue ACRBCD9-HF251-25UG
Fournisseur:  ACROBIOSYSTEMS
Frequently Bought Together