FITC-Labeled Human CD4, Fc Tag (CD4-HF255) is expressed from human 293 cells (HEK293). It contains AA Lys 26 - Pro 396 (Accession P 01730). It is the FITC labeled form of Human CD4, Fc Tag (CD4-H5259). 1 * 25 µG
N° de catalogue ACRBCD4-HF255-25UG
Fournisseur: ACROBIOSYSTEMS
Frequently Bought Together