FITC-Labeled Human CD4, Fc Tag (CD4-HF255) is expressed from human 293 cells (HEK293). It contains AA Lys 26 - Pro 396 (Accession P 01730). It is the FITC labeled form of Human CD4, Fc Tag (CD4-H5259). 1 * 200 µG
Catalog # ACRBCD4-HF255200UG
Anbieter: ACROBIOSYSTEMS
Frequently Bought Together