FITC-Labeled Human CD4, His Tag (CD4-HF2H7) is expressed from human 293 cells (HEK293). It contains AA Lys 26 - Trp 390 (Accession AAH25782). It is the FITC labeled form of Human CD4, His Tag (LE3-H5228). 1 * 200 µG
N° de catalogue ACRBCD4-HF2H7200UG
Fournisseur: ACROBIOSYSTEMS
Frequently Bought Together