Stereo microscopes, ergonomic, Mantis
Stereo microscopes, ergonomic, Mantis
MCO objective 2×
N° de catalogue VISIMCO-002
Fournisseur:   VISION ENGINEERING
Stereo microscopes, ergonomic, Mantis
N° de catalogue VISIMCO-002
Fournisseur:  VISION ENGINEERING
Stereo microscopes, ergonomic, Mantis
Stereo microscopes, ergonomic, Mantis
MCO objective 2×
Prix
Demander un prix
Connexion Pour la disponibilité
1 * 1 ST
Product Details & Documents
Superior imaging for a wide range of inspection and rework tasks.
  • Patented optical technology for fatigue-free viewing and superb image quality
  • Wide range of magnification options to 20x
  • Long working distances; large depths of field
  • Shadow-free true colour LED illumination
  • Choice of table stand or articulated arm
Mantis Compact excels in the low magnification range for inspection or manipulation tasks where bench magnifiers have traditionally been used. Mantis Compact has a small footprint and low nvestment cost giving a superb price/performance ratio.
Mantis Elite has enhanced optical performance, including higher magnification, a large field of view plus long working distances, making it a perfect alternative to traditional stereo microscopes for a wide range of inspection, preparation and manipulation tasks requiring hand-eye co-ordination.
Ordering information: Objective lenses sold separately.
Documents
Spécifications
  • Désignation:
    MCO objective 2×