ESC (Easy Surface Checking) swabs
ESC swab D/E Neutralising Broth, 100 tubes×10 ml
N° de catalogue 85605.
Fournisseur: LIOFILCHEM
Aller à
Product Details & Documents
The ESC (Easy Surface Checking) swab range includes a new generation of sampling devices for microbiological examination of surfaces, especially recommended for food and pharmaceutical industries.
- Pre-filled swab device
- Rayon bud
- Wide range of diluents
- Leak proof closure
- Transport and storage at room temperature
Certifications: Swabs conform to international standards (ISO 18593, ISO 17604).
Spécifications
- Désignation:ESC swab D/E Neutralising Broth, 100 tubes×10 ml
- Seq:04