Objectives for Rebel Microscopes
Plan achromat oil phase, N.A. 1,25, 100×, 0,2 mm
Catalog # ECHLRMAG1003
Supplier: DISCOVER ECHO
Jump to:
Product Details & Documents
Documents
Specifications
- Grossissement:100×
- Désignation:Plan achromat oil phase, N.A. 1,25
- Seq:06
- Distance de travail:0,2 mm