BUCHER Agilent Microplates unique 5 cavity polypropylene reservoir, 4 control wells (cols 23-24 rows A-H, I-P) of 2.6ml each and 145 ml main, 384 pyramid base geometries, 30.6mm height, case of 2
Référence MSPP201300-100
Fournisseur: MARKET SOURCE PART PROCESS
Frequently Bought Together