Saisie de commande
Switzerland
Lien pour commandesContactUsLinkComponent
Transfection reagents, FluoMag™

Transfection reagents, FluoMag™

FluoMag™-P transfection reagent corresponding to PolyMag (DNA transfection reagent), 100 µl
Catalog # OZBIFP10100
Supplier:   OZ BIOSCIENCES
Transfection reagents, FluoMag™
Catalog # OZBIFP10100
Supplier:  OZ BIOSCIENCES
CAS Number:  
Transfection reagents, FluoMag™
Transfection reagents, FluoMag™

FluoMag™-P transfection reagent corresponding to PolyMag (DNA transfection reagent), 100 µl

CAS Number:  
Price
SFr. 143.90
Log in for availability
1 * 100 µl
Product Details & Documents

FluoMag™ reagents are fluorescently-labeled magnetic nanoparticles for Magnetofection™ applications. FluoMag™ transfection reagents are as efficient as their non labelled counterparts and allow visualisation of the nanoparticles in vitro during experiments.

  • Advantages of Magnetofection™ technology
  • Easy cell labelling and tracking
  • Non toxic
  • Serum compatible

Store at +4 °C and protect from the light.

Ordering information: This reagent needs to be used with a magnetic plate, available separately.

Delivery information: Shipped at room temperature.

Specifications

Specifications

  • Cdt:
    100 µl
  • Désignation:
    FluoMag™-P transfection reagent corresponding to PolyMag (DNA transfection reagent)

Specifications

Accessories

Product Information

  • Catalog #
    Désignation
    Availability
    Price
  • OZBIMF10000
    Super magnetic plate, 8×12 cm
    Log in for availability
    1 * 1 ST - SFr. 363.68

Product Information

  • Catalog #
    Désignation
    Availability
    Price
  • OZBIMF14000
    Mega magnetic plate, 20×26 cm
    Log in for availability
    1 * 1 ST - SFr. 711.20

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.