Saisie de commande
Switzerland
Lien pour commandesContactUsLinkComponent
 

Transfection reagents, FluoMag™

FluoMag™-N transfection reagent corresponding to NeuroMag (neuron transfection), 100 µl
  OZBIFN10100
Transfection reagents, FluoMag™
  OZBIFN10100
 :  
Transfection reagents, FluoMag™
Transfection reagents, FluoMag™

FluoMag™-N transfection reagent corresponding to NeuroMag (neuron transfection), 100 µl

 :  
 
SFr. 104.72
   
1 * 100 µl
 

FluoMag™ reagents are fluorescently-labeled magnetic nanoparticles for Magnetofection™ applications. FluoMag™ transfection reagents are as efficient as their non labelled counterparts and allow visualisation of the nanoparticles in vitro during experiments.

  • Advantages of Magnetofection™ technology
  • Easy cell labelling and tracking
  • Non toxic
  • Serum compatible

Store at +4 °C and protect from the light.

 : This reagent needs to be used with a magnetic plate, available separately.

 : Shipped at room temperature.

 

 

  • Cdt:
    100 µl
  • Désignation:
    FluoMag™-N transfection reagent corresponding to NeuroMag (neuron transfection)
  • Cat. No.:
    OZBIFN10100
  • Spécificité cellulaire:
    Neurons

 

Accessories

 

  •  
    Désignation
     
     
  • OZBIMF10000
    Super magnetic plate, 8×12 cm
       
    1 * 1 ST - SFr. 363.68

 

  •  
    Désignation
     
     
  • OZBIMF14000
    Mega magnetic plate, 20×26 cm
       
    1 * 1 ST - SFr. 711.20

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.