Colour coded inserts, CapInsert™
pourpre
Katalognr. SIMPM957L
Anbieter: Simport Scientific
Direkt zu
Product Details & Documents
These colour coded inserts could be applied to the tube closure, which eliminates jeopardising the integrity of your sample by transferring it to another vial.
- Manually press fit and locks into place on top of closure
Technische Daten
- Couleur:pourpre
- Seq:03